Learn More
R&D Systems™ Human CEACAM-1/CD66a (NP_001703) VersaClone cDNA
Carcinoembryonic antigen-related cell adhesion molecule 1
Brand:  R&D Systems™ RDC0951
Encompass
Product Code. 15700945
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
The Human CEACAM-1/CD66a (NP_001703) VersaClone cDNA clone contains the complete ORF for CEACAM1/CD66a, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CEACAM1/CD66a insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
CEACAM1/CD66a
634
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
NP_001703
antigen CD66, BGP1, BGP-1, BGP1BGPBiliary glycoprotein 1, BGPI, Biliary Glycoprotein 1, biliary glycoprotein adhesion molecule, carcinoembryonic antigen-related cell adhesion molecule 1, carcinoembryonic antigen-related cell adhesion molecule 1 (biliaryglycoprotein), CD66a, CD66a antigen, Cea-1, CEACAM1, Hv-1, Hv-2, MHVR
Human
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human CEACAM-1/CD66a (NP_001703) VersaClone cDNA
For Reasearch Use Only; Not for Use in Humans