Learn More
Invitrogen™ Human OR10Q1 Control Fragment Recombinant Protein
Recombinant Protein
Brand: Invitrogen™ RP100610
Description
Highest antigen sequence indentity to the following orthologs: Mouse (69%), Rat (69%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61385 (PA5-61385. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Odorant receptor.
Specifications
Q8NGQ4 | |
Blocking Assay, Control | |
219960 | |
100 μL | |
olfactory receptor 10Q1; olfactory receptor family 10 subfamily Q member 1; olfactory receptor OR11-233; olfactory receptor, family 10, subfamily Q, member 1; OR10Q1; OR11-233 | |
OR10Q1 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human OR10Q1 Control Fragment | |
RUO | |
OR10Q1 | |
Unconjugated | |
Recombinant | |
GKLVFNQSDPTEFVFRAFTTATEFQV | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.