Learn More
Lonza™ Human Stem Cell Nucleofector™ Kit 2
For efficient transfection of primary human stem cells in the Nucleofector™ I/II/2b Device.
Brand: Lonza™ VPH-5022
This item is not returnable.
View return policy
Description
- High transfection efficiencies for primary cells.
- Cells maintain functionality post transfection.
- Applications:
- Transfection of cell numbers in the range of 2 × 105 to 2 × 107 cells.
- Low-throughput transfection in a single cuvette.
Specifications
Clear | |
Nucleofector™ I/II/2b Device | |
2.25 mL Nucleofector™ Solution for Human Stem Cells, 0.5 mL Supplement 1, 30 μg pmaxGFP™ Vector (0.5 μg/μL in 10 mM Tris pH 8.0), 25 x Aluminum cuvettes (100 μL), 25 x Single-use pipettes | |
8 |
pmaxGFP™ | |
Efficient transfection of primary human stem cells | |
25 | |
1 Kit |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
Product Title
Lonza™ Human Stem Cell Nucleofector™ Kit 2 > 2.25 mL Nucleofector™ Solution for Human Stem Cells, 0.5 mL Supplement 1, 30 μg pmaxGFP™ Vector (0.5 μg/μL in 10 mM Tris pH 8.0), 25 x Aluminum cuvettes (100 μL), 25 x Single-use pipettes
Spot an opportunity for improvement?Share a Content Correction