missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech HumanKine™ recombinant human IL-8 protein
HumanKine™ recombinant protein
80.00 EUR - 2087.00 EUR
Specifications
Accession Number | P10145 |
---|---|
Formulation | 1 x PBS |
Gene ID (Entrez) | 3576 |
Molecular Weight (g/mol) | 12 kDa |
Source | HEK293 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17660077
|
Proteintech
HZ-1318-1000UG |
1000 μg |
2087.00 EUR
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17670077
|
Proteintech
HZ-1318-100UG |
100 μg |
417.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17680077
|
Proteintech
HZ-1318-10UG |
10 μg |
80.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-8 is a chemoattractant cytokine that signals through the CXCR1 and CXCR2 receptors to attract and activate neutrophils to sites of inflammation. It is secreted by multiple types of cells, including monocytes, neutrophils, tumor, epithelial, endothelial, and fibroblasts. IL-8 signaling activates downstream pathways that promote cell survival, proliferation, and cell invasiveness. As such, IL-8 has been linked to the promotion of angiogenesis, immunosuppressive cell recruitment, and EMT transition in several cancer types. Recent studies also indicate that IL-8 can serve as a marker for COVID-19 disease progression (PMID: 22235381, 18980965, 26548401, 33584733).Specifications
P10145 | |
3576 | |
HEK293 | |
<0.1 EU/μg | |
Recombinant | |
Lyophilized |
1 x PBS | |
12 kDa | |
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.) | |
Human | |
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). | |
>95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Proteintech HumanKine™ recombinant human IL-8 protein