Learn More
Cytiva HyClone™ Phosphate Buffered Saline (PBS)
Product Code. 10793004
Click to view available options
Quantity:
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
Concentration:
1 X
10 X
Unit Size:
1000mL
500mL
Pack of 6
Product Code. 10793004
Brand: Cytiva SH30256.LS
Buffer cell culture growth with PBS
  • QC-tested solution
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Phosphate Buffered Saline (PBS)
Sterilization Method 0.1 μm Sterile-filtered
Without Additives No Calcium, No Magnesium, No Phenol Red
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.2
Quantity 6 x 1000 mL