Learn More
Lonza™ AccuGENE™ 10% SDS (Monosodium Salt)
Ideal for a wide range of molecular biology applications
Brand:  Lonza™ 51213
 View more versions of this product
Product Code. 11508025
  • 198.00 EUR / Each
Explore more special offers
Description
Description
  • Ready-to-use
  • Prepared with 18megohm water
  • Filtered using 0.2μ filter and filled into sterile bottles
  • Manufactured according to strict quality control standards to ensure consistency
  • Guaranteed DNase, RNase and protease-free
  • Customized solutions are available to meet individual needs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yes
Ideal for a wide range of molecular biology applications
Yes
10% SDS (w/v), monosodium salt
10% SDS (Monosodium Salt)
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lonza™ AccuGENE™ 10% SDS (Monosodium Salt) > 10% SDS (Disodium salt); 500mL
For research or manufacturing use only.