Learn More
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents
An automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples, when paired with a KingFisher purification system.
Brand:  Applied Biosystems™ A66600
 View more versions of this product
Product Code. 30054388
  • 316.00 EUR / 0.40mL
Explore more special offers
Description
Show More
Specifications
Specifications
Store at room temperature
Magnetic Bead
KingFisher™ Duo Prime, Flex, or Apex purification systems
Genomic DNA, Total RNA, miRNA
Sequential DNase I
Room Temperature
Reagent
Whole Blood, Bone Marrow
Molecular applications, including qPCR and sequencing
MagMAX™
0.4 mL
Flexible
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2969076 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA66600
Lot Number3134697 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA66600
Lot Number3141176 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA66600
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents > Sequential DNase I, 0.4 mL
For Research Use Only. Not for use in diagnostic procedures.