Learn More
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents
An automated solution for the sequential isolation of both high-quality genomic DNA and total RNA from whole blood or bone marrow samples, when paired with a KingFisher purification system.
Brand:  Applied Biosystems™ A66592
 View more versions of this product
Product Code. 30054784
  • 253.00 EUR / 120mL
Description
Show More
Specifications
Specifications
Store at room temperature
Magnetic Bead
KingFisher™ Duo Prime, Flex, or Apex purification systems
Genomic DNA, Total RNA, miRNA
RNA Binding Solution
Room Temperature
Reagent
Whole Blood, Bone Marrow
Molecular applications, including qPCR and sequencing
MagMAX™
120 mL
Flexible
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2971883 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberA66592
Lot Number3140288 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberA66592
Product Title
Applied Biosystems™ MagMAX™ Sequential DNA/RNA Kit and Reagents > Sequential RNA Binding Solution, 120 mL