missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Human CD117 (c-Kit) Positive Selection Kit

Designed for the magnetic separation of CD117+ cells by positive selection. It has been optimized for the isolation of CD117+ cells from normal human peripheral blood cells.

Brand:  Invitrogen™ 8802-6870-74

534.69 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


Product Code. 15551267

  • 802.00 EUR / 200 tests
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Contains Sodium azide

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price


Includes:
  • MagniSort Anti-Human CD117 (c-Kit) Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads B
This item is not returnable. View return policy

Description

Description

  • CD117+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD117+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD117, clone YB5.B8, and Anti-Human CD34
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD117+
200 tests
Human
Positive Selection Kit
200 tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3101154 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number8802-6870-74
Lot Number2410140 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8802-6870-74
Lot Number2478078 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8802-6870-74
Lot Number2532159 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8802-6870-74
Lot Number2729933 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number8802-6870-74
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ MagniSort™ Human CD117 (c-Kit) Positive Selection Kit > MagniSort Human CD117 (c-Kit) Positive Selection Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.