Learn More
Invitrogen™ MagniSort™ Mouse CD45 Depletion Kit
Designed for the magnetic depletion of mouse CD45+ cells from mouse spleens, bone marrow or digested lung tissue. It uses a biotinylated Anti-Mouse CD45 antibody and streptavidin-coated magnetic beads.
Brand: Invitrogen™ 8804-6864-74
524.02 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
Contains Sodium azide
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes:
- MagniSort Anti-Mouse CD45 Biotin A : 200 tests, 20μL/test
- MagniSort Negative Selection Beads B
Description
- CD45+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the CD45- cells can be separated from CD45+ cells by decanting
- After depletion, the purity of enriched CD45- cells can be verified by staining with Anti-Mouse CD45, clone 30-F11

Specifications
CD45+ | |
200 tests | |
Mouse |
Cell Depletion Kit | |
200 tests |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.