Learn More
MBD1 Antibody (100B272.1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 8 publications
Brand: Bio-Techne NB100-56537SS
Description
MBD1 Monoclonal specifically detects MBD1 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Specifications
MBD1 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 2-4 ug/ml, Chromatin Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 25798578), Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 24464130), Immunoprecipitation 1:10-1:500. Use reported in scientific literature (PMID 15327775), Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24464130), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, KnockDown Validated | |
CXXC-type zinc finger protein 3, methyl-CpG binding domain protein 1, methyl-CpG binding domain protein 1 isoform PCM1, methyl-CpG-binding domain protein 1, Methyl-CpG-binding protein MBD1, PCM1CXXC3RFT, Protein containing methyl-CpG-binding domain 1, the regulator of fibroblast growth factor 2 (FGF-2) transcription | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
There are many isoforms of human MBD1 ranging from 503 (∼55 kDa) to 655 (∼71 kDa) amino acids in length. Researchers are encouraged to use the provided immunogen sequence to determine if this antibody is suitable for testing a specific isoform of interest. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown | |
100B272.1 | |
Unconjugated | |
Q9UIS9 | |
MBD1 | |
This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1; GenBank no ref NP_056671.2 | |
0.025 mg | |
Chromatin Research | |
4152 | |
Human, Mouse | |
Purified |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only