Learn More
MDA 19, MedChemExpress
MedChemExpress MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.
Brand:  Medchem Express HY-15451
Encompass_Preferred
 View more versions of this product
Product Code. 16429117
  • 91.80 EUR / 5mg
Description
MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.
Specifications
Specifications
MDA 19
1048973-47-2
5 mg
C21H23N3O2
DMSO : 14.29 mg/mL (40.90 mM; Need ultrasonic)
349.43
98.22%
Solid
Yellow
Research
Neuroscience-Neuromodulation
O=C(N/N=C1C(N(CCCCCC)C2=C\1C=CC=C2)=O)C3=CC=CC=C3
349.43
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MDA 19, MedChemExpress