Learn More
MDA 19, MedChemExpress
MedChemExpress MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.
Brand: Medchem Express HY-15451
Description
MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.Specifications
MDA 19 | |
1048973-47-2 | |
5 mg | |
C21H23N3O2 | |
DMSO : 14.29 mg/mL (40.90 mM; Need ultrasonic) | |
349.43 | |
98.22% |
Solid | |
Yellow | |
Research | |
Neuroscience-Neuromodulation | |
O=C(N/N=C1C(N(CCCCCC)C2=C\1C=CC=C2)=O)C3=CC=CC=C3 | |
349.43 | |
Research |
Product Suggestions
Customers who viewed this item also viewed