Learn More
MDA 19, MedChemExpress
MedChemExpress MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.

91.80 EUR - 535.95 EUR
Specifications
Chemical Name or Material | MDA 19 |
---|---|
Physical Form | Solid |
CAS | 1048973-47-2 |
Color | Yellow |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16486528
![]() |
Medchem Express
HY-15451 |
10 mg |
133.65 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16442627
![]() |
Medchem Express
HY-15451 |
50 mg |
535.95 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16429117
![]() |
Medchem Express
HY-15451 |
5 mg |
91.80 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDA 19 is a potent and selective agonist of human cannabinoid receptor 2 (CB2), with a Ki of 43.3 nM. MDA 19 has antiallodynic effects in a rat model of neuropathic pain and does not affect rat locomotor activity.Specifications
MDA 19 | |
1048973-47-2 | |
Research | |
Neuroscience-Neuromodulation | |
O=C(N/N=C1C(N(CCCCCC)C2=C\1C=CC=C2)=O)C3=CC=CC=C3 | |
349.43 | |
Research |
Solid | |
Yellow | |
C21H23N3O2 | |
DMSO : 14.29 mg/mL (40.90 mM; Need ultrasonic) | |
349.43 | |
98.22% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.