missing translation for 'onlineSavingsMsg'
Learn More
Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals Product Code.: 11467417

Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals

Product Code. 11467417
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11467417

Brand: Thermo Scientific Alfa Aesar A15582.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1117-71-1
Molecular Formula C5H7BrO2
Molecular Weight (g/mol) 179.01
MDL Number MFCD00000246,MFCD09998308
InChI Key RWIKCBHOVNDESJ-NSCUHMNNSA-N
Synonym methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl esterShow MoreShow Less
PubChem CID 5369175
IUPAC Name methyl (E)-4-bromobut-2-enoate
SMILES COC(=O)\C=C\CBr

Specifications

Density 1.524
Boiling Point 83°C to 85°C (13 mmHg)
Flash Point 91°C (195°F)
Linear Formula BrCH2CH=CHCO2CH3
Refractive Index 1.501
Quantity 50 g
UN Number UN1760
Beilstein 1745755
Solubility Information Slightly soluble in water.
Formula Weight 179.01
Percent Purity 85%
Grade Technical
Chemical Name or Material Methyl 4-bromocrotonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10234388 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberA15582.30,A15582.18,A15582.09
Lot NumberF3095A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15582.30,A15582.18,A15582.09
Lot Number10234388 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15582.30,A15582.18,A15582.09
Lot NumberE8953B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15582.30,A15582.18,A15582.09
Lot NumberG9659A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15582.30,A15582.18,A15582.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.