Learn More
MF498, MedChemExpress
MedChemExpress MF498 is a novel and selective E prostanoid receptor 4 (EP4 receptor) antagonist, displayed strong binding affinity for the EP4 receptor with Ki of 0.7 nM.

POA - 264.60 EUR
Specifications
Chemical Name or Material | MF498 |
---|---|
CAS | 915191-42-3 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C32H33N3O7S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16476867
![]() |
Medchem Express
HY-10794 |
5 mg |
153.90 EUR
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16411099
![]() |
Medchem Express
HY-10794 |
1 mg |
POA
1mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16404168
![]() |
Medchem Express
HY-10794 |
10 mg |
264.60 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MF498 is a novel and selective E prostanoid receptor 4 (EP4 receptor) antagonist, displayed strong binding affinity for the EP4 receptor with Ki of 0.7 nM.Specifications
MF498 | |
Yellow | |
C32H33N3O7S | |
O=C1N(C2=C(C)C=C(CS(NC(CC3=CC=CC=C3OC)=O)(=O)=O)C=C2)CC4=C(OCC)C5=NC=CC=C5C(OCC)=C41 | |
603.69 | |
Research | |
Metabolism-protein/nucleotide metabolism |
915191-42-3 | |
Research | |
DMSO : ≥ 31 mg/mL (51.35 mM) | |
603.69 | |
98.15% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.