Learn More
MP Biomedicals™ Casein
Low Trace Metals
Brand: MP Biomedicals™ 0296012825
This item is not returnable.
View return policy
Description
Precipitated from bovine milk by hydrochloric acid rather than lactic acid to remove most of the trace metals usually present in casein. Used in different animal diets.
Specifications
Animal Diet | |
Powder | |
11.33 kg | |
15°C to 30°C |
Protein 95%, Moisture 5%, Fat 1.5%, Lactose 0.2%, Ash 2.1%, pH (10% Solution) 4.6 to 5.1, Free Acids 0.2% | |
Used in different animal diet, blocking agen in immunochemistry | |
Rat |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
MP Biomedicals™ Casein > Casein, 25 lbs
Spot an opportunity for improvement?Share a Content Correction