missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas

Activity 50 u/mg protein, Porcine Pancreas, Milky Solution

Brand:  MP Biomedicals™ 0210061750


 View more versions of this product

Product Code. 11497720

  • 227.00 EUR / 50mg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2X
Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
Porcine pancreas
Elastase
DFP, Elastinal, alpha-2-macroglobulin
Milky
4°C
50 mg
≈25000
Solution
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas > 50mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.