missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas

Activity 50 u/mg protein, Porcine Pancreas, Milky Solution

66.80 EUR - 392.00 EUR

Specifications

Concentration 2X
Color Milky
For Use With (Application) Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
Content And Storage 4°C
Source Porcine pancreas
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11487720
View Documents
MP Biomedicals™
0210061725
25 mg
122.00 EUR
25mg
Please to purchase this item. Need a web account? Register with us today!
11477720
View Documents
MP Biomedicals™
0210061710
10 mg
66.80 EUR
10mg
Please to purchase this item. Need a web account? Register with us today!
11497720
View Documents
MP Biomedicals™
0210061750
50 mg
227.00 EUR
50mg
Please to purchase this item. Need a web account? Register with us today!
11407730
View Documents
MP Biomedicals™
0210061780
100 mg
392.00 EUR
100mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.

Specifications

Specifications

2X
Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes
Porcine pancreas
≈25000
Solution
Milky
4°C
Elastase
DFP, Elastinal, alpha-2-macroglobulin
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.