Learn More
MP Biomedicals™ Pepsin
From porcine stomach mucosa
Brand: MP Biomedicals™ 0219536701
Description
Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.

Specifications
Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies | |
Porcine Stomach Mucosa | |
Lyophilized | |
Pepsin |
1 to 3.5 | |
0°C | |
1 g |
Safety and Handling
- Pepsin
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Pepsin
Your input is important to us. Please complete this form to provide feedback related to the content on this product.