missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Pepsin
From porcine stomach mucosa
103.00 EUR - 801.00 EUR
Spezifikation
For Use With (Application) | Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies |
---|---|
pH Range | 1 to 3.5 |
Sample Type | Porcine Stomach Mucosa |
Content And Storage | 0°C |
Form | Lyophilized |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | ||||||
11455322
|
MP Biomedicals™
0219536705 |
5 g |
441.00 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11445322
|
MP Biomedicals™
0219536701 |
1 g |
103.00 EUR
1g |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
11465322
|
MP Biomedicals™
0219536710 |
10 g |
801.00 EUR
10g |
Estimated Shipment: 02-05-2025
Log in to see stock. |
||||||
Beschreibung
Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.
Spezifikation
Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies | |
Porcine Stomach Mucosa | |
Lyophilized |
1 to 3.5 | |
0°C | |
Pepsin |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
MP Biomedicals™ Pepsin