Learn More
MP Biomedicals Sodium Deficient Diet, Modified
Rat
Brand:  MP Biomedicals 0290290210
 View more versions of this product
Product Code. 11485672
  • 810.00 EUR / 10kg
Explore more special offers
Specifications
Sodium Deficient Diet
Pellet
10,000 g
2°C to 8°C
Casein Purified High Nitrogen 20%, Sucrose 67%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 7%, Sodium Free Salt Mixture (Hartcroft and Eisenstein) 4% Plus MP Vitamin Diet Fortification Mixture
Powder
Rat
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
MP Biomedicals Sodium Deficient Diet, Modified > 10kg; Pelleted