missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Sodium Deficient Diet, Modified

Rat

810.00 EUR - 2905.00 EUR

Specifications

Product Type Sodium Deficient Diet
Components Casein Purified High Nitrogen 20%, Sucrose 67%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 7%, Sodium Free Salt Mixture (Hartcroft and Eisenstein) 4% Plus MP Vitamin Diet Fortification Mixture
Format Pellet
Species Rat
Storage Requirements 2°C to 8°C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11405682
View Documents
MP Biomedicals
0290290250
50,000 g
2905.00 EUR
50kg
Please to purchase this item. Need a web account? Register with us today!
11495672
View Documents
MP Biomedicals
0290290220
20,000 g
1335.00 EUR
20kg
Please to purchase this item. Need a web account? Register with us today!
11485672
View Documents
MP Biomedicals
0290290210
10,000 g
810.00 EUR
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Sodium Deficient Diet
Pellet
2°C to 8°C
Casein Purified High Nitrogen 20%, Sucrose 67%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 7%, Sodium Free Salt Mixture (Hartcroft and Eisenstein) 4% Plus MP Vitamin Diet Fortification Mixture
Rat
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals Sodium Deficient Diet, Modified

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.