Learn More
MP Biomedicals Sodium Deficient Diet, Modified
Rat
Brand: MP Biomedicals 0290290210
This item is not returnable.
View return policy
Description
Specifications
Sodium Deficient Diet | |
Pellet | |
10,000 g | |
2°C to 8°C |
Casein Purified High Nitrogen 20%, Sucrose 67%, Alphacel, Non-nutritive Bulk 2%, Corn Oil 7%, Sodium Free Salt Mixture (Hartcroft and Eisenstein) 4% Plus MP Vitamin Diet Fortification Mixture | |
Powder | |
Rat |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
MP Biomedicals Sodium Deficient Diet, Modified > 10kg; Pelleted
Spot an opportunity for improvement?Share a Content Correction