missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP3/NALP3 Antibody (25N10E9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-03948
This item is not returnable.
View return policy
Description
NLRP3/NALP3 Monoclonal specifically detects NLRP3/NALP3 in Human, Mouse samples. It is validated for Western Blot, Simple Western.
Specifications
NLRP3/NALP3 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 2-5 ug/ml, Simple Western 1:50 | |
AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, NLRP3 monoclonal, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
The epitope that 25N10E9 binds to the NOD region of NLRP3/NALP3 has not been mapped. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot | |
25N10E9 | |
Unconjugated | |
NP_001073289 | |
NLRP3 | |
A recombinant protein corresponding to the NOD domain of murine NLRP3/NALP3 Antibody (25N10E9) (amino acids 216-385) was used as the immunogen for this antibody. | |
0.1 mg | |
Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
114548 | |
Human, Mouse | |
Purified |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NLRP3/NALP3 Antibody (25N10E9) - BSA Free, Novus Biologicals™ > 0.1mg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction