missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP3/NALP3 Antibody (25N10E9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
211.00 EUR - 415.00 EUR
Specifications
Antigen | NLRP3/NALP3 |
---|---|
Clone | 25N10E9 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 2-5 ug/ml, Simple Western 1:50 |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18078776
|
Bio-Techne
NBP2-03948SS |
0.025 mg |
211.00 EUR
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789033
|
Novus Biologicals
NBP2-03948 |
0.1 mg |
415.00 EUR
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NLRP3/NALP3 Monoclonal specifically detects NLRP3/NALP3 in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
NLRP3/NALP3 | |
1.0 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
NP_001073289 | |
114548 | |
A recombinant protein corresponding to the NOD domain of murine NLRP3/NALP3 Antibody (25N10E9) (amino acids 216-385) was used as the immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
25N10E9 | |
Western Blot 2-5 ug/ml, Simple Western 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, NLRP3 monoclonal, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
NLRP3 | |
IgG2b κ | |
Protein G purified | |
The epitope that 25N10E9 binds to the NOD region of NLRP3/NALP3 has not been mapped. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
NLRP3/NALP3 Antibody (25N10E9) - BSA Free, Novus Biologicals™