Learn More
Thermo Scientific™ Orion™ Pro Star Series Meter Accessories
Select Thermo Scientific™ Orion™ Pro Star series meter accessories to support your Orion Pro Star PH211, EC212, DO213, PI214, PC215, PD216, PI217 and MP218 bench meters.
Brand:  Thermo Scientific™ PSTAR-UMS
 View more versions of this product
Product Code. 30245784
  • 62.00 EUR / 1 set
Explore more special offers
Description
Show More
Specifications
Specifications
USB Mouse
Tested to be compatible with Orion Pro Star meters and allows the meter to be operated with a remote device, removing the need to touch the display to operate the meter, which can be helpful in environments where sample carryover may damage the meter
1 USB mouse
Orion Pro Star series bench electrochemistry meters
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Orion™ Pro Star Series Meter Accessories > Pro Star USB mouse