Learn More
Hughes Outdoor Self-Draining Safety Shower with Eye/Face Wash
This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates.
Brand: Hughes HSHDSD18SS
This item is not returnable.
View return policy
Description
- This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates
- The design allows standing water to drain away from the standpipe to ensure the delivery of tepid water
- Fitted with foot panel as standard
- The valve is situated directly above the water inlet; the bleed pipe is fitted above this so standing water can drain into the ground preventing stagnant water overheating from solar radiation and removing the risk of further injury to the operator
- Available with galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Push Lever, Foot Treadle | |
Stainless Steel | |
Safety Shower |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Product Title
Hughes Outdoor Self-Draining Safety Shower with Eye/Face Wash > Safety Shower; Material: Stainless Steel
Spot an opportunity for improvement?Share a Content Correction