Learn More
Cytiva Peristaltic Pump
Single-channel peristaltic pump combines reproducible flow rates with simple operation. Cytiva Peristaltic Pump functions either as a standalone unit, or interfaced with programmable fraction collectors.
Brand: Cytiva 18-1110-91
3340.20 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Features- Low pulsation flow from 0.6 to 500 mL/h,
- Automatic pressure plate for convenient tubing changes
- Maximum and reverse flow functions
Specifications
2 each silicone tubing 1mm, 2.1mm, 3.1mm i.d., 6 tubing connectors, Allen key 3mm, Flow rate guide, O-ring fuse and fuse, Support rod/P-1 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13