Learn More
KD Scientific Plastic Syringe
Economical and disposable, available in sizes ranging from 1 cc to 50 cc, Luer Slip (LS) or Luer Lock (LL) tip
34.30 EUR - 184.00 EUR
Specifications
Needle Gauge | No Needle Included |
---|---|
Latex Free | Yes |
Product Type | Syringe |
Disposable | Single-use |
For Use With (Equipment) | KD Scientific Syringe Pumps |
Product Code | Brand | Volume (Metric) | Connection Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Connection Type | Price | Quantity & Availability | ||||||
15174194
|
KD Scientific
780850 |
1 mL | Tuberculin Luer Slip |
34.30 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15194194
|
KD Scientific
780851 |
3 mL | Luer Lock |
55.80 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15104204
|
KD Scientific
780852 |
5 mL | Luer Lock |
55.80 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15134204
|
KD Scientific
780853 |
10 mL | Luer Lock |
60.45 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15114204
|
KD Scientific
780854 |
20 mL | Luer Lock |
73.20 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15124204
|
KD Scientific
780855 |
30 mL | Luer Lock | N/A | N/A | N/A | |||||
15154204
|
KD Scientific
780857 |
3 mL | Luer Slip |
60.45 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15164204
|
KD Scientific
780858 |
5 mL | Luer Slip |
57.20 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15174204
|
KD Scientific
780859 |
10 mL | Luer Slip |
129.00 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15184204
|
KD Scientific
780860 |
20 mL | Luer Slip |
130.00 EUR
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
15194204
|
KD Scientific
780861 |
30 mL | Luer Slip | N/A | N/A | N/A | |||||
Description
- A new line of disposable plastic sterile syringes for all scientific applications
- Norm-ject syringes are the ideal solution for any situation
- Their unique two-part system is latex free and contains no silicone lubricant or rubber
Specifications
No Needle Included | |
Syringe | |
KD Scientific Syringe Pumps | |
Sterile |
Yes | |
Single-use | |
Yes |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
KD Scientific Plastic Syringe