Learn More
SCHOTT AG Lighting and Imaging™ Polarization Filter Set for Ring Light
This Polarization Filter Set for Ring Light provides contrast with polarized light by using a polarizing and analyzing filter within the beam path. Assists in providing contrast with Stereo Microscopy to help identify inner structure materials or metal particles within others.
353.94 EUR - 460.00 EUR
Specifications
Type | Filter |
---|---|
For Use With (Application) | Stereo Microscopy Illumination |
For Use With (Equipment) | Fiber Optic Ringlight |
Includes | Polarizer and analyzer |
Description
- Polarization Filter Set for fiber optic Ring Light.
- Fits inside diameters of 58 or 66 mm.
- Includes polarizer and analyzer.
- For use with fiber optic Ring Light.
Specifications
Filter | |
Stereo Microscopy Illumination | |
Fiber Optic Ringlight | |
Polarizer and analyzer |