Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand:  Quickfit XA42
60.99 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11996124
  • 90.95 EUR / Pack of 2
Estimated Shipment: 30-05-2025
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Specifications
19/26
Borosilicate Glass
29/32
Useful for the connection of dissimilar sized ground Glass joints
Expansion adapter
Documents
Product Information
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    18
    Hours
    0
    Minutes
    59
    Seconds
    58
Product Content Correction
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 19/26; Socket size: 29/32