Learn More
Memmert™ Rack for shaking device for 45 test tubes
Rack for shaking device for 25 test tubes
159.00 EUR
Specifications
Type | Rack for shaking device for 45 test tubes 32 mm |
---|---|
Diameter (Metric) | 32 mm |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17150315
|
Memmert™
WTB15B48820 |
WTB15 |
159.00 EUR
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
17150345
|
Memmert™
WTB24B48820 |
WTB24 |
159.00 EUR
Each |
N/A | ||||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Rack for shaking device for test tubes
- Accessory
Specifications
Rack for shaking device for 45 test tubes 32 mm | |
32 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Memmert™ Rack for shaking device for 45 test tubes