missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Raf-1 Protein, Active

Active, N-terminal GST-tagged recombinant human Raf-1 residues 306-end, containing the mutations Y340D and Y341D, for use in Kinase Assays.

Encompass_Preferred

525.15 EUR - 4008.15 EUR

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 65 kDa
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17300164
Encompass_Preferred
View Documents
Eurofins DiscoverX
14-352
10 μg
525.15 EUR
10µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17310164
Encompass_Preferred
View Documents
Eurofins DiscoverX
14-352M
250 μg
4008.15 EUR
250µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Raf-1 is the cellular homolog of viral raf gene (v-raf). The encoded protein is a MAP kinase kinase kinase (MAP3K), which functions downstream of the Ras family of membrane associated GTPases to which it binds directly. Once activated, the cellular RAF1 protein can phosphorylate to activate the dual specificity protein kinases MEK1 and MEK2, which in turn phosphorylate to activate the serine/threonine specific protein kinases, ERK1 and ERK2. Activated ERKs are pleiotropic effectors of cell physiology and play an important role in the control of gene expression involved in the cell division cycle, apoptosis, cell differentiation and cell migration. Mutations in this gene are associated with Noonan syndrome 5 and LEOPARD syndrome 2.

EMBL X03484

Specifications

Specifications

mg/mL of enzyme in 50 mM Tris/HCl, pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Human
N-terminal GST
Truncated
Raf-1 (truncated), active
65 kDa
Recombinant
Recombinant enzyme expressed in Sf21 insect cells
≥60%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Eurofins DiscoverX Raf-1 Protein, Active

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.