missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL1 / MGSA / NAP-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL1 / MGSA / NAP-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8650-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_032202.1 | |
CXCL1 / MGSA / NAP-3 Protein | |
Research Use Only | |
CXCL1 | |
Recombinant Protein | |
E. coli | |
NELRCQCLQT MAGIHLKNIQ SLKVLPSGPH CTQTEVIATL KNGREACLDP EAPLVQKIVQ KMLKGVPK | |
Greater than 90% as determined by SDS-PAGE. |
14825 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse CXCL1 / MGSA / NAP-3 Protein > 200μg
Spot an opportunity for improvement?Share a Content Correction