missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL1 / MGSA / NAP-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL1 / MGSA / NAP-3, was expressed in the hosts and tags indicated.
245.00 EUR - 3545.00 EUR
Specifications
Accession Number | NP_032202.1 |
---|---|
Gene ID (Entrez) | 14825 |
Name | CXCL1 / MGSA / NAP-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075761
|
enQuireBio™
QP8650-ec-10ug |
10 μg | N/A | ||||||
16015771
|
enQuireBio™
QP8650-ec-50ug |
50 μg | N/A | ||||||
16065761
|
enQuireBio™
QP8650-ec-100ug |
100 μg |
800.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16095761
|
enQuireBio™
QP8650-ec-200ug |
200 μg | N/A | ||||||
16005771
|
enQuireBio™
QP8650-ec-500ug |
500 μg | N/A | ||||||
16085761
|
enQuireBio™
QP8650-ec-1mg |
1 mg |
3545.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_032202.1 | |
CXCL1 / MGSA / NAP-3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
Tris-based buffer, 50% glycerol |
14825 | |
Research Use Only | |
CXCL1 | |
Recombinant Protein | |
NELRCQCLQT MAGIHLKNIQ SLKVLPSGPH CTQTEVIATL KNGREACLDP EAPLVQKIVQ KMLKGVPK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse CXCL1 / MGSA / NAP-3 Protein