missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL39 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5101390
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RPL39.
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.
Specifications
RPL39 | |
Polyclonal | |
Unconjugated | |
Rpl39 | |
2810465O16Rik; 60S ribosomal protein L39; L39; Large ribosomal subunit protein eL39; ribosomal protein L39; Rpl39; RPL39_23_1806; RPL39P42 | |
Rabbit | |
Affinity chromatography | |
RUO | |
25347, 6170, 67248 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P62891, P62892, P62893 | |
Rpl39 | |
A synthesized peptide derived from human RPL39, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPL39 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction