missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL27A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5106580
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RPL27A.
Rpl27a gene ontology annotations related to this gene include translation.
Specifications
RPL27A | |
Polyclonal | |
Unconjugated | |
RPL27A | |
60S ribosomal protein L27a; 60S ribosomal protein L27-A; 60S ribosomal protein L27a-like protein; fa93g12; FLJ43464; hypothetical protein LOC336726; L27'; L27A; L29; Large ribosomal subunit protein eL27-A; Large ribosomal subunit protein uL15; MGC10850; MGC17878; MGC87238; ORF; ORF protein; ribosomal 60S subunit protein L27A; ribosomal protein L27A; ribosomal protein L27a-like; ribosomal protein L29 homolog; RPL27; RPL27A; wu:fa93g12; YHR010W; zgc:77235 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26451, 293418, 6157 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P14115, P18445, P46776 | |
RPL27A | |
A synthesized peptide derived from human RPL27A, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 15
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPL27A Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction