missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud CG / Chromogenic C. Albicans Agar

Isolate fungi and presumptively identify Candida albicans from clinical specimens with this space saving, convinient bi-plate.

Brand:  Thermo Scientific™ PO5243E

Product Code. 10322273

  • 25.60 EUR / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Isolate fungi and presumptively identify Candida albicans from clinical specimens, with ready-to-use Thermo Scientific™ Oxoid™ Sabouraud GC / Chromogenic C albicans Agar, a single bi-plate which includes two separate formulations. The Sabouraud GC medium creates selective conditions for isolation and cultivation of fungi while Chromogenic C. albicans Agar enables the presumptive identification of Candida albicans. The convenient bi-plate format frees up incubator space.

Use Sabouraud GC / Chromogenic C. albicans Agar for isolation of fungi and presumptive identification of Candida albicans from clinical specimens.

  • Suitable for identification of pathogenic fungi from material containing large numbers of other fungi or bacteria.
  • Reliable results with Microsporum audouini, Microsporum canis, Trichophyton rubrum and Candida albicans.
  • Easy isolation and presumptive identification of albicans.
  • Ready-to-use convenience of prepared media.
  • Space saving bi-plate format.

Sabouraud GC Agar is suitable for cultivation and differentiation of fungi. The fungi maintain their typical cultural appearance thus may be readily identified according to macroscopic characters.

Chromogenic C albicans Agar is a selective differential medium for the isolation of Candida spp. And presumptive identification of C. albicans. Differentiation of C. albicans is achieved by the addition of chromogenic substrate to detect the activity of enzyme β-Glucosaminidase.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ivory/light ivory
90mm Biplate
Agar
10/Pk.
2°C to 10°C
Germany
17g ± 5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6255082 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPO5243E
Lot Number6249181 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPO5243E
Lot Number6245997 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberPO5243E
Lot Number6239812 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberPO5243E
Lot Number6237643 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberPO5243E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Sabouraud CG / Chromogenic C. Albicans Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.