missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Sephadex™ G-25
Used in affinity chromatography, protein chromatography and gel filtration chromatography. Sephadex™ G-25 is a gel filtration medium that has been used to study the acceleration of diabetic wound healing and the treatment of epithelial damage.
Brand: MP Biomedicals™ 0219525450
This item is not returnable.
View return policy
Description
- Gel filtration medium used in affinity chromatography, protein chromatography and gel filtration chromatography
- Has been used to study the acceleration of diabetic wound healing and the treatment of epithelial damage

Specifications
50 g | |
Sephadex G-25 Media | |
Store at Room Temperature (15°C to 30°C) |
100 to 150 cm/hr. | |
Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Sephadex™ G-25 > Sephadex™ G-25; Quantity: 50 g
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction