missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic heptahydrate, Honeywell Fluka™
ACS reagent, 98.0-102.0%
24.69 EUR - 843.31 EUR
Chemical Identifiers
CAS | 7782-85-6 |
---|---|
Molecular Formula | H17Na2O11P |
Molecular Weight (g/mol) | 270.08 |
MDL Number | MFCD00149180 |
InChI Key | PYLIXCKOHOHGKQ-UHFFFAOYSA-N |
Synonym | sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate |
PubChem CID | 6096963 |
SMILES | O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15678130
|
Honeywell-Fluka
S9390-100G |
100 g | |||||||
15618140
|
Honeywell-Fluka
S9390-500G |
500 g |
81.95 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15688130
|
Honeywell-Fluka
S9390-1KG |
1 kg |
138.80 EUR
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15698130
|
Honeywell-Fluka
S9390-2.5KG |
2.5 kg |
330.00 EUR
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15628140
|
Honeywell-Fluka
S9390-6X500G |
6 x 500 g |
385.61 EUR
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15608140
|
Honeywell-Fluka
S9390-4X2.5KG |
4 x 2.5 kg | |||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7782-85-6 | |
270.08 | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
6096963 |
H17Na2O11P | |
MFCD00149180 | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O |
Specifications
7782-85-6 | |
Na2HPO4 · 7H2O | |
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate | |
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O | |
270.08 | |
268.07g/mol | |
1.68g/mL at 25°C; Vapor Density: 4.9 (vs air) |
H17Na2O11P | |
MFCD00149180 | |
PYLIXCKOHOHGKQ-UHFFFAOYSA-N | |
disodium;hydrogen phosphate;heptahydrate | |
6096963 | |
98 to 102% | |
Sodium phosphate dibasic heptahydrate |