missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SRSF1 Monoclonal Antibody (96), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 324500
This item is not returnable.
View return policy
Description
SRSF1 Monoclonal antibody specifically detects SRSF1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications
SRSF1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q07955, Q6PDM2 | |
SRSF1 | |
Recombinant SF2/ASF protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
96 | |
Unconjugated | |
SRSF1 | |
1110054N12Rik; 5730507C05Rik; 6330415C05Rik; AI482334; Alternative-splicing factor 1; ASF; ASF/SF2; ASF-1; AW491331; OK/SW-cl.3; pre-mRNA-splicing factor SF2, P33 subunit; pre-mRNA-splicing factor SRp30a; serine and arginine rich splicing factor 1; serine/arginine-rich splicing factor 1; SF2; SF2p33; SFRS1; splicing factor 2; splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1; SRp30a; Srsf1 | |
Mouse | |
Protein A | |
RUO | |
110809, 6426, 689890 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SRSF1 Monoclonal Antibody (96), Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction