Learn More
BINDER™ Stacking Adapter
Stacking adapter to stack two devices.
Brand: BINDER™ 9051-0056
This item is not returnable.
View return policy
Description
- Used to safely stack two identical devices
- For use with KB 65, KB 130, KB PRO 130, KBF 130, or KBF PRO 130 models
Specifications
KB 65 | |
Plastic | |
Stacking Adapter |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BINDER™ Stacking Adapter > For use with KB 65
Spot an opportunity for improvement?Share a Content Correction