Learn More
BINDER™ Stacking Adapter
Stacking adapter to stack two devices.
453.00 EUR - 495.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
30238228
|
BINDER™
9051-0048 |
KB 130, KB PRO 130, KBF 130, KBF PRO 130 |
495.00 EUR
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
30238371
|
BINDER™
9051-0056 |
KB 65 |
453.00 EUR
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
||||||
Description
- Used to safely stack two identical devices
- For use with KB 65, KB 130, KB PRO 130, KBF 130, or KBF PRO 130 models
Specifications
Plastic | |
Stacking Adapter |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BINDER™ Stacking Adapter