Learn More
Thermo Scientific™ Sterilin™ Pipette Bulbs
Transfer liquids safely using glass pipettes and these natural rubber pipette bulbs.
Brand: Thermo Scientific™ TET0102
267.49 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Transfer liquids safely using glass pipettes and these natural rubber Sterilin™ Pipette Bulbs.
- Ideal for use with glass Pasteur pipettes
- Manufactured from natural rubber
- Volume drawn up: 0.22 cm3
Ordering Information
- Not available in all geographic regions
Specifications
Pipette bulbs; No. 10 Teat (Case of 100) | |
Glass Pasteur Pipette | |
Natural Rubber | |
Ideal for use with glass Pasteur pipettes |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M561220 | Certificate of Compliance | 28/03/2025 | TET0102 |
M560130 | Certificate of Compliance | 10/03/2025 | TET0102 |
M556040 | Certificate of Compliance | 11/02/2025 | TET0102 |
M554200 | Certificate of Compliance | 10/12/2024 | TET0102 |
M553320 | Certificate of Compliance | 10/12/2024 | TET0102 |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30