Learn More
BRAND™ Straight Burette Stopcock for Burette
Straight Burette Stopcock for Burette includes an interchangeable glass key.
Brand:  BRAND™ 81815
 View more versions of this product
Product Code. 10365471
Description
Description
  • Stopcock housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable borosilicate glass key with retention device
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Burette Stopcock
Borosilicate Glass (3.3)
2 to 10 mL Burette
Glass Key
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Straight Burette Stopcock for Burette > 2 to 10 mL Burette Cap; Clear Borosilicate Glass 3.3