Learn More
Applied Biosystems™ TaqMan™ Gene Expression Control Kit
The Applied Biosystems™ TaqMan™ Gene Expression Control Kit is designed to detect and quantify eukaryotic 18S ribosomal RNA (rRNA) and human telomerase reverse transcriptase (hTERT) in human Raji cDNA on the QuantStudio™ 5 Real-Time PCR System.
Brand:  Applied Biosystems™ A29349
Product Code. 15930263
  • 502.00 EUR / 250 reactions
Explore more special offers
Description
Description
Gene-specific, ready-to-use TaqMan gene expression assays provide you with a fast, simple way to measure changes in gene expression in real-time PCR experiments.
Features of the TaqMan Gene Expression Control Kit include:
• Simple—includes pre-formulated assays with our universal cycling conditions
• Easy to use—just add to your real-time PCR experiment with no additional steps
• Robust—generate reproducible quantitative gene expression data
Ready-to-use gene expression solution
The TaqMan Gene Expression Control Kit contains TaqMan gene expression assays for eukaryotic 18S rRNA and hTERT and a sample of human Raji cDNA for use in TaqMan-based relative quantitation on the QuantStudio 5 Real-Time PCR System.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store in freezer (-5°C to -30°C).
Gene Expression Control Kit
TaqMan™
Approved for shipment on Wet or Dry Ice
18 S rRNA & hTERT
250 Reactions
Tube
FAM
250 reactions
Human
Gene Expression
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ TaqMan™ Gene Expression Control Kit > Store in freezer (-5 to -30°C).
For Research Use Only. Not for use in diagnostic procedures.