Learn More
TES-1025, MedChemExpress
MedChemExpress TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
Brand: Medchem Express HY-111365
Description
TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.Specifications
TES-1025 | |
Yellow | |
C18H13N3O3S2 | |
DMSO : 100 mg/mL (260.80 mM; Need warming) | |
383.44 | |
95.11% | |
Solid |
1883602-21-8 | |
Research | |
10 mg | |
O=C1C(C#N)=C(C2=CC=CS2)N=C(SCC3=CC=CC(CC(O)=O)=C3)N1 | |
383.44 | |
Research | |
Metabolism-protein/nucleotide metabolism |