Learn More
Thermo Scientific™ CL2 Accessories
For use with Swinging-bucket rotor
Brand: Thermo Scientific™ 003230F
This item is not returnable.
View return policy
Description
- Metal
- Sealed
- Holds 50mL test tube
- Fits 825S rotor
- Use with Falcon corning centrifuges
Specifications
825S Rotor for Falcon/Corning centrifuge |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Thermo Scientific™ CL2 Accessories > Sealed Dome Shield, 50ml
Spot an opportunity for improvement?Share a Content Correction