Learn More
TPP™ Flat Sided Cell Culture Flasks
Manufactured from polystyrene for crystal clear transparency. TPP™ Tissue Culture Flasks provide enhanced cell adhesion and growth through opto-mechanical tissue culture treated growth area.
Brand: TPP™ 90076
103.50 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Manufactured from polystyrene for crystal clear transparency. TPP™ Tissue Culture Flasks provide enhanced cell adhesion and growth through opto-mechanical tissue culture treated growth area.
- Surface activated, uniform flat growth surface
- Geometry of the neck reduces risk of media slop over
- Stable, slip-free stacking of multiple TPP flasks
- Highly visible bilateral marking area
- White and accurate volume graduations for optic control of the filling volume
- Geometry of the neck guarantees total access of TPP pipettes, TPP scrappers and the TPP spatula
- No dead corners = 100 % cell retrieval
- Excellent access of pipettes, tissue scrapers and tissue spatula
- Marking area with lines on both sides
- Increased overflow height reduces the danger of slopover of media
Specifications
Polyethylene | |
Polystyrene | |
Tissue Culture Treated | |
75 cm2 |
65 mL | |
Filter cap with PTFE membrane; pore size 0.22μm | |
Filter Screw Cap | |
8 to 22 mL |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZL402626 | Certificate of Analysis | 12/02/2025 | 90076 |