Learn More
JULABO™ Ultra-Low Refrigerated-Heating Circulator, ME Series
Ultra-low Refrigerated Circulator with 2-stage cascade-cooling. Powerful circulating pump systems and high heating and cooling capacities guarantee short heat-up and cool-down times. Equipped with an improved insulation to help avoid ice-formation.
Brand: JULABO™
This item is not returnable.
View return policy
Description
- VFD COMFORT DISPLAY
- Keypad for setpoints, warning/safety values and menu functions
- PID3 cascade temperature control
- ATC3 3-Point-Calibration
- Pt100 External sensor connection for measurement and control
- SMART PUMP, electronically adjustable pump stages
- Early warning system for low liquid level (DBGM 203 06 059.8)
- Adjustable high temperature cut-out, visible via display
- RS232 interface for online communication
- Integrated programmer for 10 program steps
- Active Cooling Control
- Easily removed venting grill allows for easy cleaning
- Equipped with handles or castors for an easy transportation
- Visual liquid level display
Specifications
5 to 40 °C | |
6.5 L | |
-81°C to +100°C | |
1.3 kW | |
F81-ME | |
±0.02°C | |
58 x 50 x 88 cm | |
15 cm | |
13 cm | |
16 cm | |
86 kg | |
50/60 Hz |
Ultra-Low Refrigerated-Heating Circulator | |
0.45 kW at 20°C | |
VFD | |
2 each barbed fittings for tubing 8 and 10 mm I.D. (pump connections M10x1 female) | |
11-16 L/min | |
16 x 13 x 15 cm | |
58 cm | |
50 cm | |
88 cm | |
86 kg | |
230 V |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
JULABO™ Ultra-Low Refrigerated-Heating Circulator, ME Series > Range: -81 to 100°C
Spot an opportunity for improvement?Share a Content Correction